missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2/CD102 Antibody (041), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89424PCP
This item is not returnable.
View return policy
Description
ICAM-2/CD102 Monoclonal antibody specifically detects ICAM-2/CD102 in Human samples. It is validated for ELISA
Specifications
| ICAM-2/CD102 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 041 | |
| ELISA | |
| CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ICAM-2/CD102 (Accession#: NP_000864.2; Met 1-Gln 223). | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3384 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction