missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2/CD102 Antibody (B-T1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14603APC
This item is not returnable.
View return policy
Description
ICAM-2/CD102 Monoclonal antibody specifically detects ICAM-2/CD102 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Functional Assay
Specifications
| ICAM-2/CD102 | |
| Monoclonal | |
| APC | |
| CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
| ICAM-2 transfected CHO cell line | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3384 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunoprecipitation, Functional Assay | |
| B-T1 | |
| PBS | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction