Learn More
ICAM-2/CD102 Antibody (B-T1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-14603CL1
Product Code. 30152458
  • € 855.00 / 0.10mL
Description
ICAM-2/CD102 Monoclonal antibody specifically detects ICAM-2/CD102 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ICAM-2/CD102
Monoclonal
CoraFluor 1
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2
ICAM-2 transfected CHO cell line
0.1 mL
Adaptive Immunity, Immunology
3384
Store at 4°C in the dark. Do not freeze.
IgG1 κ
Flow Cytometry, ELISA, Immunoprecipitation, Functional Assay
B-T1
PBS
Mouse
Affinity purified