missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2/CD102 Antibody (B-T1), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-66624PECY55
This item is not returnable.
View return policy
Description
ICAM-2/CD102 Monoclonal specifically detects ICAM-2/CD102 in Human samples. It is validated for Flow Cytometry.
Specifications
| ICAM-2/CD102 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| ICAM2 | |
| ICAM-2 transfected CHO cells | |
| 0.1 mL | |
| Primary | |
| Recognizes the human CD102 cell surface molecule, a 55-65kD glycoprotein also known as ICAM-2. NB100-66624 inhibits cell adhesion and T cell activation and also recognizes soluble ICAM-2. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| B-T1 | |
| Flow Cytometry | |
| CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
| Mouse | |
| Protein A or G purified | |
| Adaptive Immunity, Immunology | |
| 3384.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction