missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-2/CD102 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-32236CL1
871.25 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
ICAM-2/CD102 Polyclonal antibody specifically detects ICAM-2/CD102 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ICAM-2/CD102 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Intercellular Adhesion Molecule 2 using the numbering given in entry NP_001093256.1 (GeneID 3384). | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3384 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction