Learn More
ICAM-2/CD102 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-32236R
Product Code. 18628269
  • € 633.00 / 0.10mL
Description
ICAM-2/CD102 Polyclonal antibody specifically detects ICAM-2/CD102 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ICAM-2/CD102
50mM Sodium Borate
Rabbit
0.1 mL
IgG
Polyclonal
CD102, CD102 antigen, ICAM-2, intercellular adhesion molecule 2
Immunogen affinity purified
3384