missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICAM-2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580702
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human ICAM-2/CD102.
ICAM-2 is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. ICAM-2 may play a role in lymphocyte recirculation by blocking LFA-1-dependent cell adhesion. It mediates adhesive interactions important for antigen-specific immune response, NK-cell mediated clearance, lymphocyte recirculation, and other cellular interactions important for immune response and surveillance. Several transcript variants encoding the same protein have been found for this gene. Diseases associated with ICAM2 include Colon Carcinoma In Situ.
Specifications
| ICAM-2 | |
| Polyclonal | |
| Unconjugated | |
| ICAM2 | |
| CD102; Icam2; ICAM-2; Intercellular adhesion molecule 2; Ly-60; Lymphocyte function-associated AG-1 counter-receptor; RP23-418O11.10-001 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 3384 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| 5.0 mg/mL | |
| PBS with no preservative | |
| P13598 | |
| ICAM2 | |
| Recombinant Human ICAM-2/CD102 protein (Met1-Gln223). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction