missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAM-3/CD50 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00
Specifications
| Antigen | ICAM-3/CD50 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ICAM-3/CD50 Polyclonal specifically detects ICAM-3/CD50 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| ICAM-3/CD50 | |
| Polyclonal | |
| Rabbit | |
| Cellular Markers, Immunology | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 3385 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AGGSLFVNCSTDCPSSEKIALETSLSKELVASGMGWAAFNLSNVTGNSRILCSVYCNGSQIT | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| CD50, CD50 antigen, CDW50CDw50, ICAM-3, ICAM-Rintercellular adhesion molecule-3, intercellular adhesion molecule 3 | |
| ICAM3 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title