missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ICAM-4 Monoclonal Antibody (729632)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561947
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes the Landsteiner-Wiener (LW) blood group antigen(s) that belongs to the immunoglobulin (Ig) superfamily, and that shares similarity with the intercellular adhesion molecule (ICAM) protein family. This ICAM protein contains 2 Ig-like C2-type domains and binds to the leukocyte adhesion LFA-1 protein. The molecular basis of the LW(A)/LW(B) blood group antigens is a single aa variation at position 100; Gln-100=LW(A) and Arg-100=LW(B). Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications
| ICAM-4 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q14773 | |
| ICAM4 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ICAM-4, Ala23-Ala240. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 729632 | |
| Unconjugated | |
| ICAM4 | |
| 1810015M19Rik; CD242; CD242 antigen; ICAM4; ICAM-4; Intercellular adhesion molecule 4; intercellular adhesion molecule 4 (Landsteiner-Wiener blood group); intercellular adhesion molecule 4 (LW blood group); intercellular adhesion molecule 4 isoform 3 anitgen LWa; intercellular adhesion molecule 4, Landsteiner-Wiener blood group; Landsteiner-Wiener blood group antigen a; Landsteiner-Wiener blood group glycoprotein; LW; LW blood group protein; LW glycoprotein | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3386 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction