Learn More
Invitrogen™ ICAM-5 Precursor Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103906
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ICAM5.
The protein encoded by this gene is a member of the intercellular adhesion molecule family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. This protein is expressed on the surface of telencephalic neurons and displays two types of adhesion activity, homophilic binding between neurons and heterophilic binding between neurons and leukocytes. It may be a critical component in neuron-microglial cell interactions in the course of normal development or as part of neurodegenerative diseases.
Specifications
| ICAM-5 Precursor | |
| Polyclonal | |
| Unconjugated | |
| ICAM5 | |
| CD50; Icam3; ICAM-3; Icam5; ICAM-5; intercellular adhesion molecule 3; intercellular adhesion molecule 5; intercellular adhesion molecule 5, telencephalin; telencephalin; TLCN; TLN | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 15898, 313785, 7087 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q60625, Q9UMF0 | |
| ICAM5 | |
| A synthesized peptide derived from human ICAM5(Accession Q9UMF0), corresponding to amino acid residues G889-A924. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |