Learn More
ICAM3 Mouse anti-Human, Clone: ICAM3/1019, Abnova™
Mouse monoclonal antibody raised against full length recombinant human ICAM3.
Brand: Abnova MAB13316.100ug
Description
The protein encoded by this gene is a member of the intercellular adhesion molecule (ICAM) family. All ICAM proteins are type I transmembrane glycoproteins, contain 2-9 immunoglobulin-like C2-type domains, and bind to the leukocyte adhesion LFA-1 protein. This protein is constitutively and abundantly expressed by all leucocytes and may be the most important ligand for LFA-1 in the initiation of the immune response. It functions not only as an adhesion molecule, but also as a potent signalling molecule. [provided by RefSeq]
Specifications
| ICAM3 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be dete | |
| CD50/CDW50/ICAM-R | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 3385 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2b κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| ICAM3/1019 | |
| Mouse monoclonal antibody raised against full length recombinant human ICAM3. | |
| In 10mM PBS. | |
| ICAM3 | |
| Recombinant protein corresponding to full length human ICAM3. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.