missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICAT/CTNNBIP1 Antibody (SR1112), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22453-100ul
This item is not returnable.
View return policy
Description
ICAT/CTNNBIP1 Monoclonal antibody specifically detects ICAT/CTNNBIP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
Specifications
| ICAT/CTNNBIP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| SR1112 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunoprecipitation 1:50 | |
| beta-catenin-interacting protein ICAT, catenin, beta interacting protein 1, ICATbeta-interacting protein 1, Inhibitor of beta-catenin and Tcf-4 | |
| A synthesized peptide derived from human ICAT/CTNNBIP1 (Uniprot #: Q9NSA3) | |
| 100 μg | |
| Signal Transduction, Wnt Signaling Pathway | |
| 56998 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction