missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICBP90 Binding Protein 1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-37702CL1
This item is not returnable.
View return policy
Description
ICBP90 Binding Protein 1 Polyclonal antibody specifically detects ICBP90 Binding Protein 1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ICBP90 Binding Protein 1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| C6orf107, dJ349A12.1, ICBP90, RP3-349A12.1, UHRF1 binding protein 1 | |
| This antibody maps to a region between residue 1390 to 1440 of human UHRF1 Binding Protein 1 using the numbering given in entry NP_060224.3 (GeneID 54887). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 54887 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction