missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICBP90 Binding Protein 1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37702JF646
This item is not returnable.
View return policy
Description
ICBP90 Binding Protein 1 Polyclonal antibody specifically detects ICBP90 Binding Protein 1 in Human samples. It is validated for Western Blot
Specifications
| ICBP90 Binding Protein 1 | |
| Polyclonal | |
| Western Blot |
| Western Blot | |
| Janelia Fluor 646 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction