missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICBP90 Binding Protein 1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82094MFV610
This item is not returnable.
View return policy
Description
ICBP90 Binding Protein 1 Polyclonal antibody specifically detects ICBP90 Binding Protein 1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ICBP90 Binding Protein 1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| C6orf107, dJ349A12.1, ICBP90, RP3-349A12.1, UHRF1 binding protein 1 | |
| ICBP90 Binding Protein 1 antibody was raised against a 17 amino acid synthetic peptide near the center of human ICBP90 Binding Protein 1. The immunogen is located within amino acids 890 - 940 of ICBP90 Binding Protein 1. Amino Acid Squence: DSELSPSEDRELKSDAS | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 54887 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido