Learn More
Sampling Systems™ Ice Borer
Product Code. 15889371 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
500 mm
1000 mm
Unit Size:
Each
Product Code. 15889371
Brand: Sampling Systems™ 6005A500
Only null left
Add to basket
Add to basket
Constructed with sharp cutting teeth, which allows the device to be suitable for use on frozen products such as meats. Sampling Systems™ Ice Borer is a robust and efficient sampler that is ideal for sampling ice and waxy products.
  • Suitable for ice, wax and other similar materials
  • 316 Stainless Steel
  • Rotate the sampler as it is pushed into the product
  • The sharp edge will cut into the product and the sample will be deposited in the tube
  • Push the discharge rod down the Ice Borer to remove the sample
Specifications
Product Type Ice Borer
Material Stainless Steel (316)
Autoclavable Autoclavable
For Use With (Application) Ice, wax, and other similar materials
Dimensions (L x W) 500 x 25 mm
Length (Metric) 500 mm
Width (Metric) 25 mm