missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICE1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32090JF549
This item is not returnable.
View return policy
Description
ICE1 Polyclonal antibody specifically detects ICE1 in Human samples. It is validated for Western Blot
Specifications
| ICE1 | |
| Polyclonal | |
| Western Blot | |
| Uncharacterized Protein KIAA0947 | |
| The immunogen this antibody was made to, maps to a region between residue 1582 to 1632 of human KIAA0947 using the numbering given in entry NP_056140.1 (GeneID 23379). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23379 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction