missing translation for 'onlineSavingsMsg'
Learn More

ICH/USP Oral Target Elements Standard A: Hg @ 30; As @ 15; Cd, Pb @ 5 μg/mL in 2% HNO3, VHG™

Product Code. 17927179
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 17927179

Brand: VHG Labs VHGICHUSPTELA100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICH/USP Oral Target Elements Standard A: Hg @ 30; As @ 15; Cd, Pb @ 5 μg/mL in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cadmium/Lead/Arsenic/Mercury
CAS 7440-43-9, 7439-92-1, 7440-38-2, 7439-97-6
Methods ICP/ICP-MS
Analyte or Component Names Cadmium/Lead/Arsenic/Mercury
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Cadmium 5 μg/mL/Lead 5 μg/mL/Arsenic 15 μg/mL/Mercury 30 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.