missing translation for 'onlineSavingsMsg'
Learn More

ICH/USP Oral Target Elements Standard C: Au, Ir, Os, Pd, Pt, Rh, Ru @ 100 μg/mL in 15% HCl, VHG™

Product Code. 30003107
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 30003107

Brand: VHG Labs VHGICHUSPTELC100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICH/USP Oral Target Elements Standard C: Au, Ir, Os, Pd, Pt, Rh, Ru @ 100 μg/mL in 15% HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Gold/Iridium/Osmium/Palladium/Platinum/Rhodium/Ruthenium
CAS 7440-57-5, 7439-88-5, 4-2-7440, 5-3-7440, 6-4-7440, 7440-16-6
Methods ICP/ICP-MS
Analyte or Component Names Gold/Iridium/Osmium/Palladium/Platinum/Rhodium/Ruthenium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HCl
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Gold 100 μg/mL/Iridium 100 μg/mL/Osmium 100 μg/mL/Palladium 100 μg/mL/Platinum 100 μg/mL/Rhodium 100 μg/mL/Ruthenium 100 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.