missing translation for 'onlineSavingsMsg'
Learn More

Icilin, MedChemExpress

Product Code. p-200099937
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
16440707 50 mg 50mg
16424388 10 mg 10mg
2 options
This item is not returnable. View return policy

Product Code. 16440707

Brand: Medchem Express HY11062

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Icilin (AG-3-5) is a super-agonist of the transient receptor potential M8 (TRPM8) ion channel. Icilin activates TRPM8 in EGTA in a dose-dependent manner (EC50=1.4 μM). Icilin is a 'super-cooling agent'. Icilin attenuates autoimmune neuroinflammation through modulation of the T-cell response.

Icilin (AG-3-5) is a super-agonist of the transient receptor potential M8 (TRPM8) ion channel. Icilin activates TRPM8 in EGTA in a dose-dependent manner (EC50=1.4 μM). Icilin is a 'super-cooling agent'. Icilin attenuates autoimmune neuroinflammation through modulation of the T-cell response.

Specifications

Chemical Name or Material Icilin
CAS 36945-98-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H13N3O4
Quantity 50 mg
Synonym AG-3-5
Solubility Information DMSO : ≥ 54 mg/mL (173.47 mM)
SMILES O=C1N(C2=CC=CC=C2O)CC=C(C3=CC=CC([N+]([O-])=O)=C3)N1
Molecular Weight (g/mol) 311.29
Formula Weight 311.29
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.