missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICMT Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97509MFV610
This item is not returnable.
View return policy
Description
ICMT Polyclonal antibody specifically detects ICMT in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| ICMT | |
| Polyclonal | |
| Western Blot, Immunohistochemistry-Paraffin | |
| EC 2.1.1.100, HSTE14, Isoprenylcysteine Carboxyl Methyltransferase, Isoprenylcysteine carboxylmethyltransferase, MST098, MSTP098, PCCMT, PCMT, PPMT, Prenylated Protein Carboxyl Methyltransferase, Protein-S-Isoprenylcysteine O-Methyltransferase | |
| Produced in rabbits immunized with E. coli-derived Human ICMT fragment. | |
| 0.1 mL | |
| Cell Biology | |
| 23463 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human, Cynomolgus Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction