missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS Antibody (669230), PE/Cy7, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB69751PECY7
This item is not returnable.
View return policy
Description
ICOS Monoclonal specifically detects ICOS in Human samples. It is validated for Flow Cytometry.
Specifications
| ICOS | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| ICOS | |
| Partial recombinant human ICOS protein (amino acids 21-141) [UniProt Q9Y6W8] | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 669230 | |
| Flow Cytometry | |
| Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
| Mouse | |
| Protein A or G purified | |
| Adaptive Immunity, Immunology | |
| 29851.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction