missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS Antibody (feladilimab) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28241JF525
This item is not returnable.
View return policy
Description
ICOS Monoclonal antibody specifically detects ICOS in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ICOS | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850 | |
| ICOS/ CD278 | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 29851 | |
| Store at 4°C in the dark. | |
| IgG4 |
| Flow Cytometry, ELISA, Functional Assay | |
| feladilimab | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction