missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOS Monoclonal Antibody (OTI5H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
€ 242.00 - € 540.87
Specifications
| Antigen | ICOS |
|---|---|
| Clone | OTI5H1 |
| Concentration | 1 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17517964
|
Origene Technologies
TA813215S |
30 μL |
€ 242.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17597954
|
Origene Technologies
TA813215 |
100 μL |
€ 540.87
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ICOS Monoclonal antibody specifically detects ICOS in Human samples. It is validated for Flow CytometrySpecifications
| ICOS | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Activation-inducible lymphocyte immunomediatory molecule; Ailim; CCLP; CD278; CD28 and CTLA-4-like protein; CD28-related protein 1; CRP-1; CVID1; H4; Icos; inducible costimulator; inducible T cell costimulator; inducible T cell co-stimulator; inducible T-cell costimulator; inducible T-cell co-stimulator; Ly115 | |
| Full length human recombinant protein of human ICOS (NP_036224) produced in HEK293T cell. | |
| Primary | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| ICOS |
| OTI5H1 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9Y6W8 | |
| 29851 | |
| IgG1 | |
| Protein A/G | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title