missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOSLG Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808603S
This item is not returnable.
View return policy
Description
ICOSLG Monoclonal antibody specifically detects ICOSLG in Human samples. It is validated for Western BlotSpecifications
| ICOSLG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q00403 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2959 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1B8 | |
| Unconjugated | |
| GTF2B | |
| AU044799; B7 homolog 2; B7 homologue 2; B7h; B7H2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP1; B7RP-1; BG071784; CD275; GI50; GL50; GL50-B; ICOS ligand; ICOSL; ICOS-L; ICOSLG; inducible T-cell costimulator ligand; inducible T-cell co-stimulator ligand; KIAA0653; LICOS; Ly115l; mKIAA0653; RGD1562791; transmembrane protein B7-H2 ICOS ligand | |
| Human recombinant protein fragment corresponding to amino acids 43-316 of human GTF2B produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction