missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICOSLG Monoclonal Antibody (OTI1B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808603
This item is not returnable.
View return policy
Description
CD275 is a ligand for the T-cell-specific cell surface receptor ICOS. Acts as a costimulatory signal for T-cell proliferation and cytokine secretion; induces also B-cell proliferation and differentiation into plasma cells. Could play an important role in mediating local tissue responses to inflammatory conditions, as well as in modulating the secondary immune response by co-stimulating memory T-cell function.Specifications
| ICOSLG | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75144 | |
| ICOSLG | |
| Full length human recombit protein of human ICOSLG produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1B8 | |
| Unconjugated | |
| ICOSLG | |
| B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, LICOS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23308 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction