Learn More
ICOSLG Mouse anti-Human, Clone: UMAB254, lyophilized, UltraMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies UM800144CF
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD275 is a ligand for the T-cell-specific cell surface receptor ICOS. Acts as a costimulatory signal for T-cell proliferation and cytokine secretion; induces also B-cell proliferation and differentiation into plasma cells. Could play an important role in mediating local tissue responses to inflammatory conditions, as well as in modulating the secondary immune response by co-stimulating memory T-cell function.Specifications
| ICOSLG | |
| Monoclonal | |
| Unconjugated | |
| ICOSLG | |
| B7-H2, B7H2, B7RP-1, B7RP1, CD275, GL50, ICOS-L, ICOSL, LICOS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 23308 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Peptide Array, Western Blot | |
| UMAB254 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O75144 | |
| ICOSLG | |
| Full length human recombit protein of human ICOSLG produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.