Learn More
Icotinib Hydrochloride, MedChemExpress
Product Code. 16486338
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16486338
Brand: Medchem Express HY15164
MedChemExpress Icotinib Hydrochloride (BPI-2009) is a potent and specific EGFR inhibitor with an IC50 of 5 nM; also inhibits mutant EGFRL858R, EGFRL858R/T790M, EGFRT790M and EGFRL861Q.
Icotinib Hydrochloride (BPI-2009) is a potent and specific EGFR inhibitor with an IC50 of 5 nM
Specifications
Chemical Name or Material Icotinib Hydrochloride
CAS 1204313-51-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H22ClN3O4
Quantity 10 mg
Synonym BPI-2009H
Solubility Information DMSO : 25 mg/mL (58.43 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES C#CC1=CC(NC2=C(C=C(OCCOCCOCCO3)C3=C4)C4=NC=N2)=CC=C1.[H]Cl
Molecular Weight (g/mol) 427.88
Formula Weight 427.88
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only