missing translation for 'onlineSavingsMsg'
Learn More

ICP-MS Tuning Solution: Be, Mg, Co, In, Ba, Ce, Tl, Pb, Th @ 250 ug/L in 2% HNO3, VHG™

Product Code. 17969269
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17969269 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17969269 Supplier VHG Labs Supplier No. VHGLMSTNG9500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

ICP-MS Tuning Solution: Be, Mg, Co, In, Ba, Ce, Tl, Pb, Th @ 250 ug/L in 2% HNO3
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cerium/Lead/Thorium/Barium/Beryllium/Indium/Cobalt/Magnesium/Thallium
CAS 7440-45-1, 7439-92-1, 7440-29-1, 7440-39-3, 7440-41-7, 7440-74-6
Methods ICP-MS
Analyte or Component Names Cerium/Lead/Thorium/Barium/Beryllium/Indium/Cobalt/Magnesium/Thallium
Shipping Condition Room Temperature
Solvent or Matrix Dilute HNO3
Quantity 500 mL
Concentration or Composition (by Analyte or Components) Cerium 250 μg/L/Lead 250 μg/L/Thorium 250 μg/L/Barium 250.1 μg/L/Beryllium 250.1 μg/L/Indium 250.1 μg/L/Cobalt 250.2 μg/L/Magnesium 250.2 μg/L/Thallium 250.2 μg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.