missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID1 Antibody (CL11981), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21206-100ul
This item is not returnable.
View return policy
Description
ID1 Monoclonal antibody specifically detects ID1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ID1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL11981 | |
| Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| BHLHB24, bHLHb24DNA-binding protein inhibitor ID-1, Class B basic helix-loop-helix protein 24, dJ857M17.1.2, dJ857M17.1.2 (inhibitor of DNA binding 1, dominant negative helix-loop-helixprotein), ID, inhibitor of differentiation 1, Inhibitor of DNA binding 1, inhibitor of DNA binding 1, dominant negative helix-loop-helix protein | |
| This antibody was developed against a Synthetic Peptide of P41134. Actual immunogen sequence is proprietary. | |
| 100 μg | |
| Stem Cell Markers | |
| 3397 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction