missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ID3 Monoclonal Antibody (OTI4D1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524999
This item is not returnable.
View return policy
Description
ID3 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Members of the Id family of basic helix-loop-helix (bHLH) proteins include ID1, ID2, ID3 and ID4. They are ubiquitously expressed and dimerize with members of the class A and B HLH proteins. Due to the absence of the basic region, the resulting heterodimers cannot bind DNA. The ID-type proteins thus appear to negatively regulate DNA binding of bHLH proteins. Since ID1 inhibits DNA binding of E12 and Myo D, it apparently functions to inhibit muscle-specific gene expression. Under conditions that facilitate muscle cell differentiation, the ID protein levels fall, allowing E12 and/or E47 to form heterodimers with Myo D and myogenin, which in turn activate myogenic differentiation. It has been shown that expression of each of the ID proteins is strongly dependent on growth factor activation and that reduction of ID mRNA levels by antisense oligonucleotides leads to a delayed reentry of arrested cells into the cell cycle following growth factor stimulation.
Specifications
| ID3 | |
| Monoclonal | |
| 0.82 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02535 | |
| Id3 | |
| Full length human recombinant protein of ID3 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| OTI4D1 | |
| Unconjugated | |
| Id3 | |
| 1R21; bHLHb25; class B basic helix-loop-helix protein 25; DNA-binding protein inhibitor ID-3; HEIR 1; HEIR1; HEIR-1; Helix-loop-helix protein HEIR-1; Hlh462; HLH642; ID 3; Id3; Id-3; Idb3; ID-like protein inhibitor HLH 1R21; ID-like protein inhibitor HLH 462; Inhibitor of differentiation 3; inhibitor of DNA binding 3; inhibitor of DNA binding 3, dominant negative helix-loop-helix protein; inhibitor of DNA binding 3, HLH protein | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3399 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction