missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID4 Antibody (OTI2G10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70987CL1
This item is not returnable.
View return policy
Description
ID4 Monoclonal antibody specifically detects ID4 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| ID4 | |
| Monoclonal | |
| CoraFluor 1 | |
| bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein | |
| Full length human recombinant protein of human ID4 (NP_001537) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI2G10 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 3400 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction