missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ID4 Antibody (OTI2G10), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70987V
This item is not returnable.
View return policy
Description
ID4 Monoclonal specifically detects ID4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| ID4 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ID4 | |
| Full length recombinant protein of human ID4 (NP_001537) produced in HEK293T cells. | |
| 0.1 mL | |
| 3400.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2G10 | |
| Western Blot | |
| bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction