missing translation for 'onlineSavingsMsg'
Learn More

ID4 Antibody (PCRP-ID4-1A10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20701F

Product Code. 18646954

  • € 726.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ID4 Monoclonal antibody specifically detects ID4 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ID4
Monoclonal
FITC
bHLHb27BHLHB27, Class B basic helix-loop-helix protein 27, DNA-binding protein inhibitor ID-4, IDB4, Inhibitor of DNA binding 4, inhibitor of DNA binding 4, dominant negative helix-loop-helix protein
Recombinant fragment (around aa 62-112) of human ID4 protein
0.1 mL
Primary
Human
Purified
Western Blot, ELISA, Immunoprecipitation
PCRP-ID4-1A10
PBS
Mouse
Protein A or G purified
RUO
3400
Store at 4°C in the dark.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.