Learn More
ID4 Monoclonal Antibody (OTI3H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA804983
Encompass_Preferred
 View more versions of this product
Product Code. 17544803
  • POA / 100µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
ID4 is transcription factors containing a basic helix-loop-helix (bHLH) motif regulate expression of tissue-specific genes in a number of mammalian and insect systems. DNA-binding activity of the bHLH proteins is dependent on formation of homo- and/or heterodimers. Domit-negative HLH proteins encoded by Id-related genes, such as ID4, also contain the HLH-dimerization domain but lack the DNA-binding basic domain. Consequently, Id proteins inhibit binding to DNA and transcriptional transactivation by heterodimerization with bHLH proteins.
Specifications
Specifications
ID4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P47928
ID4
Full length human recombit protein of human ID4 produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI3H1
Unconjugated
ID4
IDB4, bHLHb27
Mouse
Affinity Chromatography
RUO
3400
-20° C, Avoid Freeze/Thaw Cycles