Learn More
Invitrogen™ IDE Monoclonal Antibody (OTI4E5)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA527528
Product Code. 15987114
  • € 590.00 / 100µL
Description
IDE Monoclonal Antibody for Western Blot
This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IDE
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P14735, P35559
IDE
Human recombinant protein fragment corresponding to amino acids 800-1019 of IDE produced in E.coli.
100 μL
Primary
Human, Rat, Monkey
Antibody
IgG2b
Western Blot
OTI4E5
Unconjugated
IDE
1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603
Mouse
Affinity Chromatography
RUO
25700, 3416
-20°C, Avoid Freeze/Thaw Cycles