missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDE Mouse anti-Human, Rat, Clone: OTI4E5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811776
This item is not returnable.
View return policy
Description
This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.Specifications
| IDE | |
| Monoclonal | |
| Unconjugated | |
| IDE | |
| INSULYSIN | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 25700, 3416 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI4E5 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P14735, P35559 | |
| IDE | |
| Human recombit protein fragment corresponding to amino acids 800-1019 of human IDE produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction