missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IDE Polyclonal Antibody

Product Code. 17733234
Change view
Click to view available options
Quantity:
20 μL
200 μL
Unit Size:
200µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17733234 20 μL 20µL
17750614 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17733234 Supplier Invitrogen™ Supplier No. PA5121835

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IDE Polyclonal Antibody for Western Blot, IHC (P)

This gene may belong to a protease family responsible for intercellular peptide signalling. Though its role in the cellular processing of insulin has not yet been defined, insulin-degrading enzyme is thought to be involved in the termination of the insulin response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDE
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene IDE
Gene Accession No. P14735
Gene Alias 1300012G03Rik; 4833415K22Rik; AA675336; Abeta-degrading protease; AI507533; FLJ35968; Ide; INSDEGM; Insulin degrading enzyme; insulin protease; insulinase; Insulin-degrading enzyme; Insulysin; zgc:162603
Gene Symbols IDE
Host Species Rabbit
Immunogen Recombinant protein Insulin Degrading Enzyme. The antigen corresponds to amino acid range 753-973 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3416
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.