Learn More
Brady™ Identification Tags
Make your own tags whenever you need them. Blank Identification Tags which you can engrave yourself.
Brand: Brady™ 023210
Additional Details : Weight : 0.22450kg
Description
- The tags can also be ordered as part of the Brady ''Customize-it'' range.
Specifications
25 Pk. | |
Brass | |
Customized | |
Circle | |
-40°C to +649°C | |
0.188 in | |
Blank Valve Tags |
Customized Identification Tag | |
Brass | |
General purpose valve identification | |
1.021 mm | |
-40°F to +1200°F | |
4.78 mm | |
1.5 in. |