Learn More
IDH1 Mouse anti-Human, CoraLite 488, Clone: 2A6A2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66197-100UL
Description
The IDH1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IDH1. This antibody recognizes human antigen. The IDH1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| IDH1 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 3417 | |
| IgG1 |
| Immunofluorescence | |
| 2A6A2 | |
| Mouse | |
| Primary | |
| Human |