missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH1 Mouse, Clone: IDH1/1152, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3417-MSM2-P1
This item is not returnable.
View return policy
Specifications
| IDH1 | |
| Cytoplasmic and Nuclear | |
| IDH1/1152 | |
| Unconjugated | |
| NP_001269315 | |
| Mouse | |
| 45-47kDa | |
| Protein A/G Purified | |
| RUO | |
| 3417 | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 1mM PBS with 0.05% BSA and 0.05% azide. | |
| IDH1 | |
| Recombinant fragment (119 Amino acid residues around aa 280-420) of human IDH1 protein | |
| HepG2, HeLa, HT29 or MCF7 cells. Breast, Colon or Prostate Carcinoma. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction