missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH2, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00003418-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MAGYLRVVRSLCRASGSRPAWAPAALTAPTSQEQPRRHYADKRIKVAKPVVEMDGDEMTRIIWQFIKEKLILPHVDIQLKYFDLGLPNRDQTDDQVTIDSALATQKYSVAVKCATITPDEARVEEFKLKKMWKSPNGTIRNILGGTVFREPIICKNIPRLVPGWTKPITIGRHAHGDQYKATDFVADRAGTFKMVFTPKDGSGVKEWEVYNFPAGGVGMGMYNTDESISGFAHSCFQYAIQKKWPLYMSTKNTILKAYDGRFKDIFQEIFDKHYKTDFDKNKIWYEHRLIDDMVAQVLKSSGGFVWACKNYDGDVQSDILAQGFGSLGLMTSVLVCPDGKTIEAEAAHGTVTRHYREHQKGRPTSTNPIASIFAWTRGLEHRGKLDGNQDLIRFAQMLEKVCVETVESGAMTKDLAGCIHGLSNVKLNEHFLNTTDFLDTIKSNLDRALGRQSpecifications
| IDH2 | |
| Polyclonal | |
| No additive | |
| NM_002168.2 | |
| IDH2 | |
| IDH2 (NP_002159.2, 1 a.a. ∼ 452 a.a) full-length human protein. | |
| RUO | |
| 3418 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunoprecipitation | |
| Unconjugated | |
| isocitrate dehydrogenase 2 (NADP+), mitochondrial | |
| ICD-M/IDH/IDHM/IDP/IDPM/mNADP-IDH | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction