missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH3A Antibody (OTI2F11), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70992PCP
This item is not returnable.
View return policy
Description
IDH3A Monoclonal specifically detects IDH3A in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.
Specifications
| IDH3A | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| IDH3A | |
| Full length recombinant protein of human IDH3A (NP_005521) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG2a |
| Flow Cytometry | |
| OTI2F11 | |
| Flow Cytometry | |
| EC 1.1.1, EC 1.1.1.41, H-IDH alpha, isocitrate dehydrogenase (NAD+) alpha chain, isocitrate dehydrogenase [NAD] subunit alpha, mitochondrial, isocitrate dehydrogenase 3 (NAD+) alpha, Isocitric dehydrogenase subunit alpha, NAD(+)-specific ICDH subunit alpha, NAD(H)-specific isocitrate dehydrogenase alpha subunit, NAD+-specific ICDH | |
| Mouse | |
| Affinity Purified | |
| Stem Cell Markers | |
| 3419.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction