missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH3A Monoclonal Antibody (OTI2E9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA500912S
This item is not returnable.
View return policy
Description
IDH3A Monoclonal antibody specifically detects IDH3A in Human samples. It is validated for Flow Cytometry, ImmunocytochemistrySpecifications
| IDH3A | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96PL5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 114625 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry | |
| OTI2E9 | |
| Unconjugated | |
| ERMAP | |
| 1110003P10Rik; 1500012E04Rik; AA407078; AI316514; H-IDH alpha; Idh3a; isocitrate dehydrogenase (NAD(+)) 3 alpha; isocitrate dehydrogenase (NAD(+)) 3 catalytic subunit alpha; isocitrate dehydrogenase (NAD+) alpha chain; isocitrate dehydrogenase [NAD] subunit alpha, mitochondrial; isocitrate dehydrogenase 3 (NAD(+)) alpha; isocitrate dehydrogenase 3 (NAD+) alpha; isocitric dehydrogenase subunit alpha; NAD(+)-specific ICDH subunit alpha; NAD(H)-specific isocitrate dehydrogenase alpha subunit; NAD+-specific ICDH | |
| Full length human recombinant protein of human ERMAP produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction