missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH3G Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97338H
This item is not returnable.
View return policy
Description
IDH3G Polyclonal antibody specifically detects IDH3G in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| IDH3G | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| HRP | |
| EC 1.1.1, EC 1.1.1.41, H-IDHG, IDH-gamma, isocitrate dehydrogenase 3 (NAD+) gamma, isocitrate dehydrogenase, NAD(+)-specific, mitochondrial, gamma subunit, mitochondrial, NAD (H)-specific isocitrate dehydrogenase gamma subunit, NAD+-specific ICDH | |
| Produced in rabbits immunized with E. coli-derived Human IDH3G fragment. | |
| 0.1 mL | |
| metabolism | |
| 3421 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction