missing translation for 'onlineSavingsMsg'
Learn More

IDH3G Antibody, mFluor Violet 450 SE, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97338MFV450

Product Code. 18515293

  • € 428.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IDH3G Polyclonal antibody specifically detects IDH3G in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IDH3G
Polyclonal
Immunohistochemistry-Paraffin
EC 1.1.1, EC 1.1.1.41, H-IDHG, IDH-gamma, isocitrate dehydrogenase 3 (NAD+) gamma, isocitrate dehydrogenase, NAD(+)-specific, mitochondrial, gamma subunit, mitochondrial, NAD (H)-specific isocitrate dehydrogenase gamma subunit, NAD+-specific ICDH
Produced in rabbits immunized with E. coli-derived Human IDH3G fragment.
0.1 mL
metabolism
3421
Store at 4C in the dark.
IgG
Immunohistochemistry (Paraffin)
mFluor Violet 450 SE
50mM Sodium Borate
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.