missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IDH3G Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA585385

Product Code. 16342325

  • € 520.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keep as concentrated solution. Predicted reactivity: Mouse (98%), Rat (99%), Xenopus laevis (85%), Rabbit (98%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the gamma subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. This gene is a candidate gene for periventricular heterotopia. Several alternatively spliced transcript variants of this gene have been described, but only some of their full length natures have been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IDH3G
Polyclonal
Unconjugated
Idh3g
H-IDHG; IDH; IDH3G; IDH-gamma; iso; Isocitrate dehydrogenase [NAD] subunit gamma 1, mitochondrial; isocitrate dehydrogenase [NAD] subunit gamma, mitochondrial; isocitrate dehydrogenase 3 (NAD(+)) gamma; isocitrate dehydrogenase 3 (NAD), gamma; isocitrate dehydrogenase 3 (NAD+) gamma; isocitrate dehydrogenase 3 (NAD+), gamma; isocitrate dehydrogenase 3 gamma; isocitrate dehydrogenase 3, gamma; isocitrate dehydrogenase, NAD(+)-specific, mitochondrial, gamma subunit; isocitric dehydrogenase subunit gamma; NAD (H)-specific isocitrate dehydrogenase gamma subunit; NAD(+)-specific ICDH subunit gamma; NAD+-isocitrate dehydrogenase gamma subunit; NAD+-specific ICDH
Rabbit
Antigen affinity chromatography
RUO
3421
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P51553
Idh3g
Recombinant protein encompassing a sequence within the center region of human IDH3G. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.