missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bethyl Laboratories IDO1 Recombinant Rabbit Monoclonal Antibody (BLR040F)
Rabbit Recombinant Monoclonal Antibody
€ 133.00 - € 315.00
Specifications
| Antigen | IDO1 |
|---|---|
| Clone | BLR040F |
| Concentration | 400 μg/mL |
| Content And Storage | 4°C |
| Applications | Immunoprecipitation, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30275695
|
Bethyl Laboratories
A700040 |
100 μL |
€ 315.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276672
|
Bethyl Laboratories
A700040T |
20 μL |
€ 133.00
10µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IDO1 Recombinant Monoclonal Antibody for IP, Western Blot
IDO1 is an intracellular heme-containing enzyme that catalyzes the oxidative cleavage of the indole ring of several important regulatory molecules like tryptophan, serotonin, and melatonin. By doing this, IDO1 initiates the production of biologically active metabolites, commonly referred to as kynurenines. IDO1 is widely expressed in a variety of human tissues as well as in macrophages and dendritic cells (DCs). In inflammation, interferons (IFNs) act on specific receptors to trigger IDO1 induction. The production of IFN-gamma and induction of IDO1 represent important antimicrobial mechanisms. Degradation and depletion of tryptophan by IDO1 inhibits the growth of viruses, bacteria and parasites. Furthermore, IDO1 plays a complex and crucial role in immunoregulation during infection, pregnancy, autoimmunity, transplantation, and neoplasia.Specifications
| IDO1 | |
| 400 μg/mL | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8 to 8.6 | |
| P14902 | |
| 3620 | |
| Recombinant protein representing human Indoleamine 2, 3-dioxygenase 1 (IDO1, residues 1-403). | |
| Antibody |
| BLR040F | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Ido1 | |
| EC 1.13.11.52; IDO; IDO1; IDO-1; INDO; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; indoleamine-pyrrole 2,3-dioxygenase | |
| Ido1 | |
| Primary | |
| Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title