missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00159APC
This item is not returnable.
View return policy
Description
IDO2 Polyclonal antibody specifically detects IDO2 in Human samples. It is validated for ELISA
Specifications
| IDO2 | |
| Polyclonal | |
| ELISA | |
| EC 1.13.11.-, IDO-2, INDOL1indoleamine-pyrrole 2,3 dioxygenase-like 1, indoleamine 2,3-dioxygenase 2, indoleamine 2,3-dioxygenase-like 1 protein, Indoleamine 2,3-dioxygenase-like protein 1, Indoleamine-pyrrole 2,3-dioxygenase-like protein 1 | |
| Produced in rabbits immunized with purified, recombinant Human IDO2 (Uniprot#: Q6ZQW0-1; Met 14-Gly 420) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 169355 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction