missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO2 Antibody (LIMR patent anti-IDO2) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28269JF669
This item is not returnable.
View return policy
Description
IDO2 Monoclonal antibody specifically detects IDO2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| IDO2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 1.13.11.-, IDO-2, INDOL1indoleamine-pyrrole 2,3 dioxygenase-like 1, indoleamine 2,3-dioxygenase 2, indoleamine 2,3-dioxygenase-like 1 protein, Indoleamine 2,3-dioxygenase-like protein 1, Indoleamine-pyrrole 2,3-dioxygenase-like protein 1 | |
| IDO2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| LIMR patent anti-IDO2 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| 169355 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction